1-[4-(3-Chloropropoxy)-3-methoxyphenyl]ethan-1-one ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(4-(3-Chloropropoxy)-3-methoxyphenyl)ethanone

058635-500MG 058635-1G 058635-5G
101915-186EA 200 USD
101915-186 101915-188 101915-190
1-[4-(3-Chloropropoxy)-3-methoxyphenyl]ethan-1-one ≥95%
1-[4-(3-Chloropropoxy)-3-methoxyphenyl]ethan-1-one

Hazard: Irritant

Formula: C₁₂H₁₅ClO₃
MW: 242.7 g/mol
Melting Pt: 61…64 °C
MDL Number: MFCD05156572
CAS Number: 58113-30-7

Order Now

Specification Test Results

Melting point 61-64°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR