6-Chloro-4-(methylamino)pyridine-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Chloro-4-(methylamino)nicotinaldehyde

058630-500MG 058630-1G
101915-172EA 746.15 USD
101915-172 101915-174
6-Chloro-4-(methylamino)pyridine-3-carbaldehyde ≥95%
6-Chloro-4-(methylamino)pyridine-3-carbaldehyde

Hazard: Irritant

Formula: C₇H₇ClN₂O
MW: 170.6 g/mol
Boiling Pt: 339 °C
Melting Pt: 80…82 °C
Density: 1.357
MDL Number: MFCD14706814
CAS Number: 449811-29-4

Order Now

Specification Test Results

Melting point 80-82°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR