Benzyl-N-[2-(1H-1,2,3,4-tetrazol-5-yl)propan-2-yl]carbamate ≥95%

Supplier: Matrix Scientific

058621-500MG 058621-1G 058621-5G
101915-148EA 401.54 USD
101915-148 101915-150 101915-152
Benzyl-N-[2-(1H-1,2,3,4-tetrazol-5-yl)propan-2-yl]carbamate ≥95%
Benzyl-N-[2-(1H-1,2,3,4-tetrazol-5-yl)propan-2-yl]carbamate

Hazard: Irritant

Formula: C₁₂H₁₅N₅O₂
MW: 261.29 g/mol
Melting Pt: >100 °C (dec.)
MDL Number: MFCD18157681
CAS Number: 455875-25-9

Order Now

Specification Test Results

Melting point >100°C(dec)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR