1-Benzyl-4-hydroxypiperidine-4-carboxamide ≥95%

Supplier: Matrix Scientific

058618-500MG 058618-1G
101915-138EA 170.77 USD
101915-138 101915-140
1-Benzyl-4-hydroxypiperidine-4-carboxamide ≥95%
1-Benzyl-4-hydroxypiperidine-4-carboxamide

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂
Melting Pt: 178…180 °C
MDL Number: MFCD17947481
CAS Number: 27771-25-1

Order Now

Specification Test Results

Melting point 178-180°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR