Benzyl-N-[1-(N'-hydroxycarbamimidoyl)-1-methylethyl]carbamate ≥95%

Supplier: Matrix Scientific

058617-500MG 058617-1G 058617-5G
101915-132EA 401.54 USD
101915-132 101915-134 101915-136
Benzyl-N-[1-(N'-hydroxycarbamimidoyl)-1-methylethyl]carbamate ≥95%
Benzyl-N-[1-(N'-hydroxycarbamimidoyl)-1-methylethyl]carbamate

Hazard: Irritant

Formula: C₁₂H₁₇N₃O₃
Melting Pt: 160…162 °C
MDL Number: MFCD18157682
CAS Number: 518047-98-8

Order Now

Specification Test Results

Melting point 160-162°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR