Benzyl-N-(1-cyano-1-methylethyl)carbamate ≥95%
Supplier: Matrix Scientific
058615-1G
058615-5G
101915-126EA
500
USD
101915-126
101915-128
Benzyl-N-(1-cyano-1-methylethyl)carbamate ≥95%
Benzyl-N-(1-cyano-1-methylethyl)carbamate
Hazard: Irritant
Formula:
C₁₂H₁₄N₂O₂ MW: 218.26 g/mol Melting Pt: 100…102 °C |
MDL Number:
MFCD10698635 CAS Number: 100134-82-5 |
Specification Test Results
Melting point | 100-102°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...