2-(Azepan-1-yl)-5-bromopyridin-3-amine ≥95%

Supplier: Matrix Scientific

058614-500MG 058614-1G
101915-120EA 401.54 USD
101915-120 101915-122
2-(Azepan-1-yl)-5-bromopyridin-3-amine ≥95%
2-(Azepan-1-yl)-5-bromopyridin-3-amine

Hazard: Irritant

Formula: C₁₁H₁₆BrN₃
Melting Pt: 73…75 °C
MDL Number: MFCD14600440
CAS Number: 1226192-85-3

Order Now

Specification Test Results

Melting point 73-75°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR