2-(3,4,5-Trimethoxyphenyl)acetohydrazide ≥95%

Supplier: Matrix Scientific

058515-500MG 058515-1G
101915-008EA 375.38 USD
101915-008 101915-010
2-(3,4,5-Trimethoxyphenyl)acetohydrazide ≥95%
2-(3,4,5-Trimethoxyphenyl)acetohydrazide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₄
Melting Pt: 104…106 °C
MDL Number: MFCD17392923
CAS Number: 34547-25-6

Order Now

Specification Test Results

Melting point 104-106°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR