tert-Butyl-4-(6-methyl-2-oxo-2,3-dihydro-1H-1,3-benzodiazol-1-yl)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

058513-500MG 058513-1G
101915-000EA 346.15 USD
101915-000 101915-002
tert-Butyl-4-(6-methyl-2-oxo-2,3-dihydro-1H-1,3-benzodiazol-1-yl)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(6-methyl-2-oxo-2,3-dihydro-1H-1,3-benzodiazol-1-yl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₈H₂₅N₃O₃
MW: 331.42 g/mol
Melting Pt: 194…196 °C
MDL Number: MFCD17392914
CAS Number: 950772-99-3

Order Now

Specification Test Results

Melting point 194-196°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR