2-[(4-Fluoro-5-methyl-2-nitrophenyl)(methyl)amino]acetic acid ≥95%

Supplier: Matrix Scientific

058502-500MG 058502-1G
101914-950EA 556.92 USD
101914-950 101914-952
2-[(4-Fluoro-5-methyl-2-nitrophenyl)(methyl)amino]acetic acid ≥95%
2-[(4-Fluoro-5-methyl-2-nitrophenyl)(methyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₀H₁₁FN₂O₄
Melting Pt: 144…145 °C
MDL Number: MFCD17392918
CAS Number: 1251950-64-7

Order Now

Specification Test Results

Melting point 144-145°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR