Ethyl-6-chloro-4-(methylamino)-5-nitropyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

058500-500MG 058500-1G
101914-942EA 538.46 USD
101914-942 101914-944
Ethyl-6-chloro-4-(methylamino)-5-nitropyridine-3-carboxylate ≥95%
Ethyl-6-chloro-4-(methylamino)-5-nitropyridine-3-carboxylate

Hazard: Irritant

Formula: C₉H₁₀ClN₃O₄
MW: 259.65 g/mol
Melting Pt: 74…75 °C
MDL Number: MFCD17392924
CAS Number: 925427-22-1

Order Now

Specification Test Results

Melting point 74-75°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR