5-[(2,4-Difluorophenyl)methyl]-1,3,4-oxadiazol-2-ol ≥95%

Supplier: Matrix Scientific

058498-500MG 058498-1G
101914-936EA 546.15 USD
101914-936 101914-938
5-[(2,4-Difluorophenyl)methyl]-1,3,4-oxadiazol-2-ol ≥95%
5-[(2,4-Difluorophenyl)methyl]-1,3,4-oxadiazol-2-ol

Hazard: Irritant

Formula: C₉H₆F₂N₂O₂
Melting Pt: 103…105 °C
MDL Number: MFCD17392927
CAS Number: 1251950-58-9

Order Now

Specification Test Results

Melting point 103-105°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR