1,3-Diethyl-2-(4-fluoro-5-methyl-2-nitrophenyl)propanedioate ≥95%

Supplier: Matrix Scientific

058497-500MG 058497-1G
101914-932EA 367.69 USD
101914-932 101914-934
1,3-Diethyl-2-(4-fluoro-5-methyl-2-nitrophenyl)propanedioate ≥95%
1,3-Diethyl-2-(4-fluoro-5-methyl-2-nitrophenyl)propanedioate

Hazard: Irritant

Formula: C₁₄H₁₆FNO₆
Melting Pt: 81…83 °C
MDL Number: MFCD17392917
CAS Number: 1251950-59-0

Order Now

Specification Test Results

Melting point 81-83°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR