5-(2,4-Dichlorophenyl)-1,3,4-oxadiazol-2-ol ≥95%

Supplier: Matrix Scientific

058496-500MG 058496-1G 058496-5G
101914-926EA 176.92 USD
101914-926 101914-928 101914-930
5-(2,4-Dichlorophenyl)-1,3,4-oxadiazol-2-ol ≥95%
5-(2,4-Dichlorophenyl)-1,3,4-oxadiazol-2-ol

Hazard: Irritant

Formula: C₈H₄Cl₂N₂O₂
Melting Pt: 187…189 °C
MDL Number: MFCD17392926
CAS Number: 23767-45-5

Order Now

Specification Test Results

Melting point 187-189°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR