2-Cyano-3-(thiophen-3-yl)prop-2-enoic acid ≥95%

Supplier: Matrix Scientific

058495-500MG 058495-1G 058495-5G
101914-920EA 181.54 USD
101914-920 101914-922 101914-924
2-Cyano-3-(thiophen-3-yl)prop-2-enoic acid ≥95%
2-Cyano-3-(thiophen-3-yl)prop-2-enoic acid

Hazard: Irritant

Formula: C₈H₅NO₂S
Melting Pt: 209…211 °C
MDL Number: MFCD17392920
CAS Number: 126058-25-1

Order Now

Specification Test Results

Melting point 209-211°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR