[6-Chloro-4-(methylamino)pyridin-3-yl]methanol ≥95%

Supplier: Matrix Scientific

058492-500MG 058492-1G
101914-906EA 730.77 USD
101914-906 101914-908
[6-Chloro-4-(methylamino)pyridin-3-yl]methanol ≥95%
[6-Chloro-4-(methylamino)pyridin-3-yl]methanol

Hazard: Irritant

Formula: C₇H₉ClN₂O
Melting Pt: 172…174 °C
MDL Number: MFCD17392922
CAS Number: 449811-30-7

Order Now

Specification Test Results

Melting point 172-174°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR