3-(4-Bromothiophen-2-yl)-2-cyanoprop-2-enoic acid ≥95%

Supplier: Matrix Scientific

058490-500MG 058490-1G 058490-5G
101914-898EA 278.46 USD
101914-898 101914-900 101914-902
3-(4-Bromothiophen-2-yl)-2-cyanoprop-2-enoic acid ≥95%
3-(4-Bromothiophen-2-yl)-2-cyanoprop-2-enoic acid

Hazard: Irritant

Formula: C₈H₄BrNO₂S
Melting Pt: 230 °C (dec.)
MDL Number: MFCD17392921
CAS Number: 1087780-82-2

Order Now

Specification Test Results

Melting point 230°C(dec)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR