4-(4-Aminophenyl)morpholin-3-one ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(3-Oxomorpholino)aniline

058488-1G 058488-5G
101914-890EA 315.38 USD
101914-890 101914-892
4-(4-Aminophenyl)morpholin-3-one ≥95%
4-(4-Aminophenyl)morpholin-3-one

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₂
MW: 192.2145 g/mol
Melting Pt: 170…172 °C
MDL Number: MFCD08236742
CAS Number: 438056-69-0

Order Now

Specification Test Results

Melting point 170-172°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR