2-Chloro-4-(3-methyl-1H-pyrazol-1-yl)benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-(3-methyl-1H-pyrazol-1-yl)benzoic acid

045541-500MG 045541-1G 045541-5G
101871-378EA 492.31 USD
101871-378 101871-380 101871-382
2-Chloro-4-(3-methyl-1H-pyrazol-1-yl)benzenecarboxylic acid ≥95%
2-Chloro-4-(3-methyl-1H-pyrazol-1-yl)benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₉ClN₂O₂
MW: 236.66 g/mol
Melting Pt: 186…189 °C
MDL Number: MFCD08056672
CAS Number: 220461-68-7

Order Now

Specification Test Results

Melting point 186-189°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR