4-Bromo-5-methyl-1H-pyrazole-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

045540-500MG 045540-1G 045540-5G
101871-372EA 218.46 USD
101871-372 101871-374 101871-376
4-Bromo-5-methyl-1H-pyrazole-3-carboxylic acid ≥95%
4-Bromo-5-methyl-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₅H₅BrN₂O₂
Melting Pt: 273…276 °C (dec.)
MDL Number: MFCD08056671
CAS Number: 82231-52-5

Order Now

Specification Test Results

Melting point 273-276°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR