4-Bromo-5-methyl-1H-pyrazole-3-carbohydrazide ≥95%

Supplier: Matrix Scientific

045539-500MG 045539-1G 045539-5G
101871-366EA 556.92 USD
101871-366 101871-368 101871-370
4-Bromo-5-methyl-1H-pyrazole-3-carbohydrazide ≥95%
4-Bromo-5-methyl-1H-pyrazole-3-carbohydrazide

Hazard: Irritant

Formula: C₅H₇BrN₄O
Melting Pt: 234…237 °C
MDL Number: MFCD08056670
CAS Number: 1043909-04-1

Order Now

Specification Test Results

Melting point 234-237°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR