2-(1H-Pyrazol-3-yl)-10H-phenothiazine ≥95%

Supplier: Matrix Scientific

045537-500MG 045537-1G
101871-362EA 278.46 USD
101871-362 101871-364
2-(1H-Pyrazol-3-yl)-10H-phenothiazine ≥95%
2-(1H-Pyrazol-3-yl)-10H-phenothiazine

Hazard: Irritant

Formula: C₁₅H₁₁N₃S
Melting Pt: 225…227 °C
MDL Number: MFCD08056669
CAS Number: 63285-55-2

Order Now

Specification Test Results

Melting point 225-227°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR