3-(Dimethylamino)-1-(10H-phenothiazin-2-yl)-2-propen-1-one ≥95%
Supplier: Matrix Scientific
045535-500MG
045535-1G
045535-5G
101871-350EA
278.46
USD
101871-350
101871-352
101871-354
3-(Dimethylamino)-1-(10H-phenothiazin-2-yl)-2-propen-1-one ≥95%
3-(Dimethylamino)-1-(10H-phenothiazin-2-yl)-2-propen-1-one
Hazard: Irritant
Formula:
C₁₇H₁₆N₂OS Melting Pt: 245…247 °C |
MDL Number:
MFCD08056668 CAS Number: 63285-46-1 |
Specification Test Results
Melting point | 245-247°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...