Methyl-4-bromo-5-methyl-1H-pyrazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

045533-500MG 045533-1G 045533-5G
101871-338EA 338.46 USD
101871-338 101871-340 101871-342
Methyl-4-bromo-5-methyl-1H-pyrazole-3-carboxylate ≥95%
Methyl-4-bromo-5-methyl-1H-pyrazole-3-carboxylate

Hazard: Irritant

Formula: C₆H₇BrN₂O₂
MW: 219.04 g/mol
Melting Pt: 150…152 °C
MDL Number: MFCD08056667
CAS Number: 1232838-31-1

Order Now

Specification Test Results

Melting point 150-152°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR