4-(1,3-Benzodioxol-5-yl)benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(2H-1,3-Benzodioxol-5-yl)benzoic acid

045528-500MG 045528-1G 045528-5G
101871-310EA 300 USD
101871-310 101871-312 101871-314
4-(1,3-Benzodioxol-5-yl)benzenecarboxylic acid ≥95%
4-(1,3-Benzodioxol-5-yl)benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₀O₄
MW: 242.23 g/mol
Melting Pt: 276…278 °C
MDL Number: MFCD03426480
CAS Number: 193151-97-2

Order Now

Specification Test Results

Melting point 276-278°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR