2-Iodo-1,4-benzenediamine ≥95%

Supplier: Matrix Scientific

045527-500MG 045527-1G 045527-5G
101871-304EA 100 USD
101871-304 101871-306 101871-308
2-Iodo-1,4-benzenediamine ≥95%
2-Iodo-1,4-benzenediamine

Hazard: Irritant

Formula: C₆H₇IN₂
MW: 234.04 g/mol
Melting Pt: 109…110 °C
MDL Number: MFCD04971329
CAS Number: 69951-01-5

Order Now

Specification Test Results

Melting point 109-110°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR