2-(2,5-Dichlorophenyl)-2,3-dihydro-1,5-benzothiazepin-4(5H)-one ≥95%

Supplier: Matrix Scientific

045525-500MG 045525-1G 045525-5G
101871-292EA 375.38 USD
101871-292 101871-294 101871-296
2-(2,5-Dichlorophenyl)-2,3-dihydro-1,5-benzothiazepin-4(5H)-one ≥95%
2-(2,5-Dichlorophenyl)-2,3-dihydro-1,5-benzothiazepin-4(5H)-one

Hazard: Irritant

Formula: C₁₅H₁₁Cl₂NOS
Melting Pt: 233…235 °C
MDL Number: MFCD08056665
CAS Number: 886361-99-5

Order Now

Specification Test Results

Melting point 233-235°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR