3-(4-Bromophenyl)-2-propen-1-ol ≥95%

Supplier: Matrix Scientific

045524-500MG 045524-1G 045524-5G
101871-286EA 278.46 USD
101871-286 101871-288 101871-290
3-(4-Bromophenyl)-2-propen-1-ol ≥95%
3-(4-Bromophenyl)-2-propen-1-ol

Hazard: Irritant

Formula: C₉H₉BrO
MW: 213.08 g/mol
Melting Pt: 68…70 °C
MDL Number: MFCD08056664
CAS Number: 105515-33-1

Order Now

Specification Test Results

Melting point 68-70°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR