3-(4-Methoxyphenyl)-2-propen-1-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(4-Methoxyphenyl)prop-2-en-1-ol

045522-500MG 045522-1G 045522-5G
101871-274EA 278.46 USD
101871-274 101871-276 101871-278
3-(4-Methoxyphenyl)-2-propen-1-ol ≥95%
3-(4-Methoxyphenyl)-2-propen-1-ol

Hazard: Irritant

Formula: C₁₀H₁₂O₂
MW: 164.21 g/mol
Boiling Pt: 315 °C
Melting Pt: 76…78 °C
Density: 1.08
Flash Pt: 146.7 °C
MDL Number: MFCD00182606
CAS Number: 17581-85-0

Order Now

Specification Test Results

Melting point 76-78°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR