3'-Ethynylacetanilide ≥95%

Supplier: Matrix Scientific

Synonyms: N-(3-Ethynylphenyl)acetamide

045519-500MG 045519-1G 045519-5G
101871-262EA 192.31 USD
101871-262 101871-264 101871-266
3'-Ethynylacetanilide ≥95%
3'-Ethynylacetanilide

Hazard: Irritant

Formula: C₁₀H₉NO
MW: 159.1846 g/mol
Melting Pt: 92-93°C
MDL Number: MFCD00831583
CAS Number: 70933-58-3

Order Now

Specification Test Results

Melting point 92-93°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR