3-(Dimethylamino)-1-(3-thienyl)-2-buten-1-one ≥95%
Supplier: Matrix Scientific
045518-500MG
045518-1G
045518-5G
101871-256EA
375.38
USD
101871-256
101871-258
101871-260
3-(Dimethylamino)-1-(3-thienyl)-2-buten-1-one ≥95%
3-(Dimethylamino)-1-(3-thienyl)-2-buten-1-one
Hazard: Irritant
Formula:
C₁₀H₁₃NOS Melting Pt: 100…103 °C |
MDL Number:
MFCD06659591 CAS Number: 885949-97-3 |
Specification Test Results
Melting point | 100-103°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...