3-(2,3-Dichlorophenyl)-3-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid ≥95%

Supplier: Matrix Scientific

045515-500MG 045515-1G 045515-5G
101871-244EA 278.46 USD
101871-244 101871-246 101871-248
3-(2,3-Dichlorophenyl)-3-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid ≥95%
3-(2,3-Dichlorophenyl)-3-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)propanoic acid

Hazard: Irritant

Formula: C₁₇H₁₃Cl₂NO₃
Melting Pt: 209…212 °C
MDL Number: MFCD06659590
CAS Number: 885949-95-1

Order Now

Specification Test Results

Melting point 209-212°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR