4-[2-Amino-4-(trifluoromethyl)anilino]butanoic acid ≥95%

Supplier: Matrix Scientific

045508-500MG 045508-1G 045508-5G
101871-208EA 747.69 USD
101871-208 101871-210 101871-212
4-[2-Amino-4-(trifluoromethyl)anilino]butanoic acid ≥95%
4-[2-Amino-4-(trifluoromethyl)anilino]butanoic acid

Hazard: Irritant

Formula: C₁₁H₁₃F₃N₂O₂
Melting Pt: 144…147 °C
MDL Number: MFCD06659587
CAS Number: 106484-67-7

Order Now

Specification Test Results

Melting point 144-147°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR