2-Butyl-1,3-benzoxazol-6-amine ≥95%
Supplier: Matrix Scientific
Synonyms:
6-Amino-2-(but-1-yl)-1,3-benzoxazole
045507-500MG
045507-1G
045507-5G
101871-202EA
375.38
USD
101871-202
101871-204
101871-206
2-Butyl-1,3-benzoxazol-6-amine ≥95%
2-Butyl-1,3-benzoxazol-6-amine
Hazard: Irritant
Formula:
C₁₁H₁₄N₂O Melting Pt: 48-50°C |
CAS Number:
875850-00-3 |
Specification Test Results
Melting point | 48-50°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...