(6-Phenylimidazo[2,1-b][1,3]thiazol-5-yl)methanol ≥95%

Supplier: Matrix Scientific

045505-500MG 045505-1G 045505-5G
101871-190EA 747.69 USD
101871-190 101871-192 101871-194
(6-Phenylimidazo[2,1-b][1,3]thiazol-5-yl)methanol ≥95%
(6-Phenylimidazo[2,1-b][1,3]thiazol-5-yl)methanol

Hazard: Irritant

Formula: C₁₂H₁₀N₂OS
Melting Pt: 198…200 °C
MDL Number: MFCD02253178
CAS Number: 76919-41-0

Order Now

Specification Test Results

Melting point 198-200°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR