tert-Butyl-N-[2-(dimethylamino)-2-oxoethyl]carbamate ≥95%

Supplier: Matrix Scientific

045498-500MG 045498-1G 045498-5G
101871-156EA 278.46 USD
101871-156 101871-158 101871-160
tert-Butyl-N-[2-(dimethylamino)-2-oxoethyl]carbamate ≥95%
tert-Butyl-N-[2-(dimethylamino)-2-oxoethyl]carbamate

Hazard: Irritant

Formula: C₉H₁₈N₂O₃
MW: 202.26 g/mol
Melting Pt: 66…68 °C
MDL Number: MFCD09152695
CAS Number: 72287-76-4

Order Now

Specification Test Results

Melting point 66-68°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR