Methyl-4-(3-nitrophenyl)-2,4-dioxobutanoate ≥95%

Supplier: Matrix Scientific

045497-500MG 045497-1G 045497-5G
101871-150EA 153.85 USD
101871-150 101871-152 101871-154
Methyl-4-(3-nitrophenyl)-2,4-dioxobutanoate ≥95%
Methyl-4-(3-nitrophenyl)-2,4-dioxobutanoate

Hazard: Irritant

Formula: C₁₁H₉NO₆
Melting Pt: 111…113 °C
MDL Number: MFCD02725697
CAS Number: 345617-18-7

Order Now

Specification Test Results

Melting point 111-113°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR