4-(1,4-Dioxa-8-azaspiro[4.5]dec-8-yl)benzenecarbaldehyde ≥95%
Supplier: Matrix Scientific
045491-500MG
045491-1G
045491-5G
101871-116EA
223.08
USD
101871-116
101871-118
101871-120
4-(1,4-Dioxa-8-azaspiro[4.5]dec-8-yl)benzenecarbaldehyde ≥95%
4-(1,4-Dioxa-8-azaspiro[4.5]dec-8-yl)benzenecarbaldehyde
Hazard: Irritant
Formula:
C₁₄H₁₇NO₃ Melting Pt: 109 °C |
MDL Number:
MFCD06659581 CAS Number: 79421-40-2 |
Specification Test Results
Melting point | 109°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...