3-[4-(2-Hydroxyethyl)-3-methyl-5-oxo-2,5-dihydro-1H-pyrazol-1-yl]benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

045490-500MG 045490-1G 045490-5G
101871-110EA 181.54 USD
101871-110 101871-112 101871-114
3-[4-(2-Hydroxyethyl)-3-methyl-5-oxo-2,5-dihydro-1H-pyrazol-1-yl]benzenecarboxylic acid ≥95%
3-[4-(2-Hydroxyethyl)-3-methyl-5-oxo-2,5-dihydro-1H-pyrazol-1-yl]benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₄
Melting Pt: 102 °C
MDL Number: MFCD06659580
CAS Number: 885949-88-2

Order Now

Specification Test Results

Melting point 102°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR