3-[4-(2-Hydroxybenzoyl)-1H-pyrazol-1-yl]benzenecarboxylic acid ≥95%
Supplier: Matrix Scientific
045489-500MG
045489-1G
045489-5G
101871-104EA
375.38
USD
101871-104
101871-106
101871-108
3-[4-(2-Hydroxybenzoyl)-1H-pyrazol-1-yl]benzenecarboxylic acid ≥95%
3-[4-(2-Hydroxybenzoyl)-1H-pyrazol-1-yl]benzenecarboxylic acid
Hazard: Irritant
Formula:
C₁₇H₁₂N₂O₄ Melting Pt: 232 °C |
MDL Number:
MFCD06659579 CAS Number: 959575-06-5 |
Specification Test Results
Melting point | 232°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...