2-Butyl-5-chloro-6-nitrobenzoxazole ≥95%
Supplier: Matrix Scientific
045485-500MG
045485-1G
045485-5G
101871-080EA
278.46
USD
101871-080
101871-082
101871-084
2-Butyl-5-chloro-6-nitrobenzoxazole ≥95%
2-Butyl-5-chloro-6-nitrobenzoxazole
Hazard: Irritant
Formula:
C₁₁H₁₂N₂O₃ Melting Pt: 55…57 °C |
CAS Number:
886360-94-7 |
Specification Test Results
Melting point | 55-57°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...