2-(2-Hydroxy-1-naphthyl)acetic acid ≥95%
Supplier: Matrix Scientific
045478-500MG
045478-1G
045478-5G
101871-038EA
304.62
USD
101871-038
101871-040
101871-042
2-(2-Hydroxy-1-naphthyl)acetic acid ≥95%
2-(2-Hydroxy-1-naphthyl)acetic acid
Hazard: Irritant
Formula:
C₁₂H₁₀O₃ MW: 202.21 g/mol Melting Pt: 150 °C |
MDL Number:
MFCD00085462 CAS Number: 10441-45-9 |
Specification Test Results
Melting point | 150°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...