Imidazo[1,2-a]pyridine-6-carbohydrazide ≥95%

Supplier: Matrix Scientific

Synonyms: Imidazo[1,2-a]pyridine-6-carboxylic acid hydrazide

045477-500MG 045477-1G 045477-5G
101871-032EA 230.77 USD
101871-032 101871-034 101871-036
Imidazo[1,2-a]pyridine-6-carbohydrazide ≥95%
Imidazo[1,2-a]pyridine-6-carbohydrazide

Hazard: Irritant

Formula: C₈H₈N₄O
MW: 176.179 g/mol
Melting Pt: 219…221 °C
MDL Number: MFCD07021499
CAS Number: 886361-97-3

Order Now

Specification Test Results

Melting point 219-221°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR