Imidazo[1,2-a]pyridine-6-carboxylic acid ≥95%

Supplier: Matrix Scientific

045476-1G 045476-5G
101871-028EA 44.62 USD
101871-028 101871-030
Imidazo[1,2-a]pyridine-6-carboxylic acid ≥95%
Imidazo[1,2-a]pyridine-6-carboxylic acid

Hazard: Irritant

Formula: C₈H₆N₂O₂
Melting Pt: 305…310 °C
MDL Number: MFCD07021498
CAS Number: 139022-25-6

Order Now

Specification Test Results

Melting point 305-310°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR