5,6-Diphenyl-1,2,4-triazin-3-ol ≥95%

Supplier: Matrix Scientific

045472-500MG 045472-1G 045472-5G
101871-002EA 181.54 USD
101871-002 101871-004 101871-006
5,6-Diphenyl-1,2,4-triazin-3-ol ≥95%
5,6-Diphenyl-1,2,4-triazin-3-ol

Hazard: Irritant

Formula: C₁₅H₁₁N₃O
MW: 249.27 g/mol
Melting Pt: 224…225 °C
MDL Number: MFCD00014640
CAS Number: 4512-00-9

Order Now

Specification Test Results

Melting point 224-225°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR