Methyl-2-amino-2-(4-bromophenyl)acetate hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 2-amino-2-(4-bromophenyl)acetate hydrochloride
045468-500MG
045468-1G
045468-5G
101870-980EA
346.15
USD
101870-980
101870-982
101870-984
Methyl-2-amino-2-(4-bromophenyl)acetate hydrochloride ≥95%
Methyl-2-amino-2-(4-bromophenyl)acetate hydrochloride
Hazard: Irritant
Formula:
C₉H₁₁BrClNO₂ MW: 280.56 g/mol Melting Pt: 205…208 °C |
MDL Number:
MFCD05663802 CAS Number: 42718-20-7 |
Specification Test Results
Melting point | 205-208°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...