Ethyl-2-[(1-formyl-2-naphthyl)oxy]acetate ≥95%
Supplier: Matrix Scientific
045467-500MG
045467-1G
045467-5G
101870-974EA
181.54
USD
101870-974
101870-976
101870-978
Ethyl-2-[(1-formyl-2-naphthyl)oxy]acetate ≥95%
Ethyl-2-[(1-formyl-2-naphthyl)oxy]acetate
Hazard: Irritant
Formula:
C₁₅H₁₄O₄ Melting Pt: 90…92 °C |
MDL Number:
MFCD03001652 CAS Number: 76322-09-3 |
Specification Test Results
Melting point | 90-92°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...