2-(2-Propynyloxy)-1-naphthaldehyde ≥95%
Supplier: Matrix Scientific
045466-500MG
045466-1G
045466-5G
101870-968EA
207.69
USD
101870-968
101870-970
101870-972
2-(2-Propynyloxy)-1-naphthaldehyde ≥95%
2-(2-Propynyloxy)-1-naphthaldehyde
Hazard: Irritant
Formula:
C₁₄H₁₀O Melting Pt: 108…112 °C |
MDL Number:
MFCD02629636 CAS Number: 58758-48-8 |
Specification Test Results
Melting point | 108-112°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...