Methyl-2-(2-amino-1,3-thiazol-4-yl)acetate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-(2-aminothiazol-4-yl)acetate

045465-500MG 045465-1G 045465-5G
101870-962EA 181.54 USD
101870-962 101870-964 101870-966
Methyl-2-(2-amino-1,3-thiazol-4-yl)acetate ≥95%
Methyl-2-(2-amino-1,3-thiazol-4-yl)acetate

Hazard: Irritant

Formula: C₆H₈N₂O₂S
MW: 172.21 g/mol
Melting Pt: 119…121 °C
MDL Number: MFCD00129969
CAS Number: 64987-16-2

Order Now

Specification Test Results

Melting point 119-121°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR