1-(3-Amino-4-piperidinophenyl)-1-ethanone ≥95%

Supplier: Matrix Scientific

045461-500MG 045461-1G 045461-5G
101870-944EA 556.92 USD
101870-944 101870-946 101870-948
1-(3-Amino-4-piperidinophenyl)-1-ethanone ≥95%
1-(3-Amino-4-piperidinophenyl)-1-ethanone

Hazard: Irritant

Formula: C₁₃H₁₈N₂O
MW: 218.3 g/mol
Melting Pt: 133…136 °C
MDL Number: MFCD00116065
CAS Number: 30877-81-7

Order Now

Specification Test Results

Melting point 133-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR